Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GIPR encodes a G-protein coupled receptor for gastric inhibitory polypeptide (GIP), which was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release in the presence of elevated glucose. Mice lacking this gene exhibit higher blood glucose levels with impaired initial insulin response after oral glucose load. Defect in this gene thus may contribute to the pathogenesis of diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gastric inhibitory peptide receptor; Gastric inhibitory polypeptide receptor; GIP receptor; GIP-R; Glucose-dependent insulinotropic polypeptide receptor
Gene Aliases: GIP-R; Gippr; Gipr; Gm1081; Gm160; RATGIPPR
UniProt ID: (Mouse) Q0P543, (Rat) P43219
Entrez Gene ID: (Mouse) 381853, (Rat) 25024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support