Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAPKTAAVG PLVGAVVQGT NSTRFLVFNS KTAELLSHHK VELTQEFPKE GWVEQDPKEI LQSVYECIAR TCEKLDELNI DISNIKAVGV SNQRETTVIW DKLTGEPLYN AVVWLDLRTQ TTVEDLSKKI PGNSNFVKSK TGLPLSTYFS
GK2 (glycerol kinase 2), also known as GKP2 or GKTA, is a 553 amino acid protein that belongs to the FGGY kinase family and is involved in the pathway of glycerol degradation. Localized to the outer membrane of the mitochondrion and expressed at high levels in testis, GK2 functions to catalyze the ATPdependent conversion of glycerol to glycerol 3-phosphate. Via its catalytic activity, GK2 plays an essential role in the regulation of glycerol uptake and metabolism.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP:glycerol 3-phosphotransferase 2; GK 2; glucokinase activity, related sequence 2; glycerokinase 2; Glycerol kinase 2; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; Glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m
Gene Aliases: Gk-rs2; GK2; GKP2; Gkrs2; GKTA
UniProt ID: (Human) Q14410, (Mouse) Q9WU65
Entrez Gene ID: (Human) 2712, (Mouse) 14626, (Rat) 289481
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support