Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: NT2D1, SK-N-SH, mouse brain.
Predicted reactivity: Mouse (99%), Rat (98%), Chicken (92%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GLG1 is a protein coding gene. Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFR-1; Cysteine-rich fibroblast growth factor receptor; E-selectin ligand 1; ESL-1; Golgi apparatus protein 1; Golgi sialoglycoprotein MG-160; selectin, endothelial cell, ligand
Gene Aliases: AI593353; AW537898; CFR; CFR-1; CFR1; ESL-1; ESL1; GLG1; MG-160; MG160; Selel
UniProt ID: (Human) Q92896, (Mouse) Q61543
Entrez Gene ID: (Human) 2734, (Mouse) 20340
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support