Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Does not react with Mouse or Rat
GLG1 is a protein coding gene. Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFR-1; Cysteine-rich fibroblast growth factor receptor; E-selectin ligand 1; ESL-1; Golgi apparatus protein 1; Golgi sialoglycoprotein MG-160
Gene Aliases: CFR-1; CFR1; ESL-1; ESL1; GLG1; MG-160; MG160
UniProt ID: (Human) Q92896
Entrez Gene ID: (Human) 2734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support