Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SGCRACVLGK DFRFLGKCPK KLSEGDGAEK LRKLQPSLNS GRLLHLAMRG LGELGAQPQQ DHARWPRGSS LSECSEGDVT MANTMEEILE ESEI
The GLP2 receptor (GLP2R) is a G protein-coupled receptor superfamily member closely related to the glucagon receptor ans GLP1 receptor. Glucagon-like peptide-2 (GLP2) is a 33-amino acid proglucagon-derived peptide produced by intestinal enteroendocrine cells. Like glucagon-like peptide-1 (GLP1) and glucagon itself, it is derived from the proglucagon peptide encoded by the GCG gene. GLP2 stimulates intestinal growth and upregulates villus height in the small intestine, concomitant with increased crypt cell proliferation and decreased enterocyte apoptosis. Moreover, GLP2 prevents intestinal hypoplasia resulting from total parenteral nutrition. GLP2R, a G protein-coupled receptor superfamily member is expressed in the gut and closely related to the glucagon receptor (GCGR) and the receptor for GLP1 (GLP1R).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor GLP2R; GLP-2 receptor; GLP-2-R; GLP-2R; Glp2 receptor; Glucagon-like peptide 2 receptor; glucagon-like peptide-2 receptor
Gene Aliases: 9530092J08Rik; GLP-2; GLP2R
UniProt ID: (Human) O95838, (Rat) Q9Z0W0, (Mouse) Q5IXF8
Entrez Gene ID: (Human) 9340, (Rat) 60432, (Mouse) 93896
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support