Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RSGSAGWLD RAAGAAGAAA AAASGMESNT SSSLENLATA PVNQIQETIS DNCVVIFSKT SCSYCTMAKK LFHDMNVNYK VVELDLLEYG NQFQDALYKM TGERTVPRIF VNGTFIGGAT DTHRLHKEGK LLPLVHQCYL KKSKRKEFQ (16-164 aa encoded by BC028113)
Glutaredoxin (Grx), also known as thiol transferase, is a small heat-stable oxidoreductase. Grxs form part of the glutaredoxin system, comprising NADPH, GSH and glutathione reductase, which transfers electrons from NADPH to glutaredoxins via GSH. First discovered in E. coli as GSH-dependent hydrogen donors for ribonucleotide reductase, Grx catalyzes GSH-disulfide oxidoreductase via two redox-active cysteine residues. The active sequence (Cys-Pro-Tyr-Cys) is conserved in a variety of species. The 12-kD dithiol protein has a role in reduction of mixed disulfides in cells exposed to oxidative stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA101E13.1 (GRX2 glutaredoxin (thioltransferase) 2); glutaredoxin (thioltransferase) 2; glutaredoxin 2 (thioltransferase); Glutaredoxin-2, mitochondrial
Gene Aliases: 1700010P22Rik; AI645710; CGI-133; GLRX2; GRX2
UniProt ID: (Human) Q9NS18, (Mouse) Q923X4, (Rat) Q6AXW1
Entrez Gene ID: (Human) 51022, (Mouse) 69367, (Rat) 114022
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support