Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YVGFSNATF QSERESGDRN FAIGYYLKEK KCFPEGTDMV GILDFYFQLC SIEVTCESAS VMAATLANGG FCPITGERVL SPEAVRNTLS LMHSCGMYDF SGQFAFHVGL PAKSGVAGGI LLVVPNVMGM MCWSPPLDKM GNSVKGIHFC HDLVSLCNFH NYDNLRHFAK KLDPRREGGD QRVKSVINLL FAAYTGDVSA LRRFALSAMD MEQRDYDSRT ALHVAAAEGH VEVVKFLLEA CKVNPFPKDR WNNTPMDEAL HFGHHDVFKI LQEYQVQYTP QGDSDNGKEN QTVHKNLDGL L (370-669 aa encoded by BC038507)
Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686O15119; FLJ10358; GLS; glutaminase C; Glutaminase kidney isoform, mitochondrial; glutaminase, phosphate-activated; K-glutaminase; L-glutamine amidohydrolase; phosphate-activated glutaminase
Gene Aliases: 6330442B14; AAD20; AI314027; B230365M23Rik; GA; GAC; GAM; GLS; GLS1; Glut; KGA; KIAA0838; PAG; RATGLUT
UniProt ID: (Human) O94925, (Mouse) D3Z7P3, (Rat) P13264
Entrez Gene ID: (Pig) 399525, (Human) 2744, (Mouse) 14660, (Rat) 24398
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support