Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive Samples: SH-SY5Y, K-562, Mouse brain
Immunogen sequence: ELFSQGPRPA AIAVAGFSNW TSNFIVGMCF QYVEQLCGPY VFIIFTVLLV LFFIFTYFKV PETKGRTFDE IASGFRQGGA SQSDKTPEEL FHPLGADSQV
Facilitative glucose transporter. This isoform may be responsible for constitutive or basal glucose uptake. Has a very broad substrate specificity; can transport a wide range of aldoses including both pentoses and hexoses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: choreoathetosis/spasticity, episodic (paroxysmal choreoathetosis/spasticity); glucose transporter glut1; Glucose transporter type 1, erythrocyte/brain; GLUT-1; GT1; HepG2 glucose transporter; human T-cell leukemia virus (I and II) receptor; MGC141895; MGC141896; receptor for HTLV-1 and HTLV-2; solute carrier family 2 (facilitated glucose transporter), member 1; Solute carrier family 2, facilitated glucose transporter member 1; solute carrier family 2, member 1
Gene Aliases: CSE; DYT17; DYT18; DYT9; EIG12; GLUT; GLUT-1; GLUT1; GLUT1DS; HTLVR; PED; SDCHCN; SLC2A1
UniProt ID: (Human) P11166, (Mouse) P17809
Entrez Gene ID: (Human) 6513, (Mouse) 20525
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support