Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glucose transporter glut4; Glucose transporter type 4, insulin-responsive; GLUT-4; GT2; MGC93607; Solute carrier family 2, facilitated glucose transporter member 4
Gene Aliases: Glut-4; Glut4; Slc2a4
UniProt ID: (Mouse) P14142
Entrez Gene ID: (Mouse) 20528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support