Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CVLTAALAL QDTVSWMPYI SIVCVISYVI GHALGPSPIP ALLI (357-399 aa encoded by B C001820)
Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fructose transporter; Glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT-5; RP5-963K15.1; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; Solute carrier family 2, facilitated glucose transporter member 5; testicular tissue protein Li 81
Gene Aliases: AI526984; GLUT-5; GLUT5; SLC2A5; Slc5a
UniProt ID: (Human) P22732, (Mouse) Q9WV38
Entrez Gene ID: (Human) 6518, (Mouse) 56485
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support