Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Immunogen sequence: MWPQPRLPPR PAMSEETRQS KLAAAKKKLR EYQQRNSPGV PTGAKKKKKI KNGSNPETTT SGGCHSPEDT PKDNAATLQP SDDTVLPGGV PSPGASLTSM
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130 kDa cis-Golgi matrix protein; cis-Golgi matrix protein GM130; GM130; GM130 autoantigen; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; Golgin subfamily A member 2; Golgin-95; RP11-395P17.5; SY11 protein
Gene Aliases: AW555139; GM130; GOLGA2; mKIAA4150
UniProt ID: (Human) Q08379, (Rat) Q62839, (Mouse) Q921M4
Entrez Gene ID: (Human) 2801, (Rat) 64528, (Mouse) 99412
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support