Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSESLVVCD VAEDLVEKLR KFRFRKETNN AAIIMKIDKD KRLVVLDEEL EGISPDELKD ELPERQPRFI VYSYKYQHDD GRVSYPLCFI FSSPVGCKPE QQMMYAGSKN KLVQTAELTK VFEIRNTEDL TEEWLREKLG FFTNVNFCVS KVFMY (1-154 aa encoded by BC005359)
This protein, which is phosphorylated following phorbol ester stimulation, is important for the nervous system. It causes brain cell differentiation, stimulates neural regeneration and inhibits tumor cell proliferation. Overexpression of GMF in astrocytes has been shown to enhance brain-derived neurotrohic factor (BDNF) production. GMF expression is increased by exercise, and the protein is crucial for exercise-induction of BDNF. Through BDNF production, GMF appears to play a role in neuroprotection. In thymoma, T-cell development is maintained by GMF; being produced by the tumor cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glia maturation factor beta; glia maturation factor, beta; GMF-beta
Gene Aliases: GMF; GMFB
UniProt ID: (Human) P60983
Entrez Gene ID: (Pig) 100625819, (Human) 2764
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support