Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNPSMKQKQ EEIKENIKNS SVPRRTLKMI QPSASGSLVG RENELSAGLS KRKHRNDHLT STTSSPGVIV PESSENKNLG GVTQESFDLM IKENPSSQYW KEVAEKRRKA LYEALKENEK LHKEIEQKDN EIARLKKENK ELAEVAEHVQ YMAELIERLN GEPLDNFESL DNQEFDSEEE TVEDSLVEDS EIGTCAEGTV SSSTDAKPCI (1-209 aa encoded by BC005185)
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Geminin; Geminin GMNN; OTTHUMP00000039393; RP3 369A17.3
Gene Aliases: Gem; GMNN; MGORS6
UniProt ID: (Human) O75496
Entrez Gene ID: (Human) 51053
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support