Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides (PubMed:12009299, PubMed:12669231, PubMed:16359702, PubMed:22037469). Plays a role in modulating cellular differentiation (PubMed:12669231).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GMP reductase 2; GMPR 2; Guanosine 5'-monophosphate oxidoreductase 2; Guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog
Gene Aliases: 1810008P16Rik; 5730544D12Rik; AA959850; GMPR 2; GMPR2
UniProt ID: (Human) Q9P2T1, (Mouse) Q99L27
Entrez Gene ID: (Human) 51292, (Mouse) 105446, (Rat) 192357
Molecular Function:
reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support