Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALCNGDSKL ENAGGDLKDG HHHYEGAVVI LDAGAQYGKV IDRRVRELFV QSEIFPLETP AFAIKEQGFR AIIISGGPNS VYAEDAPWFD PAIFTIGKPV LGICYGMQMM NKVFGGTVHK KSVREDGVFN ISVDNTCSLF RGLQKEEVVL LTHGDSVDKV ADGFKVVARS GNIVAGIANE SKKLYGAQFH PEVGLTENGK VILKNFLYDI AGCSGTFTVQ NRELECIREI KERVGTSKVL VLLSGGVDST VCTALLNRAL NQEQVIAVHI DNGFMRKRES QSVEEALKKL GIQVKVINAA HSFYNGTTTL PISDEDRTPR KRISKTLNMT TSPEEKRKII GDTFVKIANE VIGEMNLKPE EVFLAQGTLR PDLIESASLV ASGKAELIKT HHNDTELIRK LREEGKVIEP LKDFHKDEVR ILGRELGLPE ELVSRHPFPG PGLAIRVICA EEPYICKDFP ETNNILKIVA DFSASVKKPH TLLQRVKACT TEEDQEKLMQ ITSLHSLNAF LLPIKTVGVQ GDCRSYSYVC GISSKDEPDW ESLIFLARLI PRMCHNVNRV VYIFGPPVKE PPTDVTPTFL TTGVLSTLRQ ADFEAHNILR ESGYAGKISQ MPVILTPLHF DRDPLQKQPS CQRSVVIRTF ITSDFMTGIP ATPGNEIPVE VVLKMVTEIK KIPGISRIMY DLTSKPPGTT EWE
Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glutamine amidotransferase; GMP synthase; GMP synthase [glutamine-hydrolyzing]; GMP synthetase; guanine monophosphate synthetase; guanine monphosphate synthetase; guanosine 5'-monophosphate synthase; MLL/GMPS fusion protein; testicular tissue protein Li 82
Gene Aliases: AA591640; AI047208; GMPS
UniProt ID: (Human) P49915, (Mouse) Q3THK7
Entrez Gene ID: (Human) 8833, (Mouse) 229363
Molecular Function: ligase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support