Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total GNA11.
Heterotrimeric guanine nucleotide-binding proteins (G proteins) consist of α, β and γ subunits and mediate the effects of hormones, neurotransmitters, chemokines and sensory stimuli. To date, over 20 known Gα subunits have been classified into four families, Gα(s), Gα(i/o), Gα(q) and Gα(12), based on structural and functional similarities (1,2). Phosphorylation of Tyr356 of Gα(q)/Gα(11) is essential for activation of the G protein, since phenylalanine substitution for Tyr356 changes the interaction of Gα with receptors and abolishes ligand-induced IP3 formation (3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G alpha-11; G-protein subunit alpha-11; GNA11; Gq class; guanine nucleotide binding protein (G protein), alpha 11 (Gq class); guanine nucleotide binding protein alpha 11; guanine nucleotide regulatory protein G alpha 11; guanine nucleotide-binding protein alpha 11 subunit; Guanine nucleotide-binding protein G(y) subunit alpha; Guanine nucleotide-binding protein subunit alpha-11
Gene Aliases: Dsk7; E430025L19; FBH; FBH2; FHH2; GA11; GNA-11; GNA11; HHC2; HYPOC2
UniProt ID: (Pig) Q2XSV9, (Human) P29992, (Mouse) P21278, (Rat) Q9JID2
Entrez Gene ID: (Pig) 733588, (Human) 2767, (Mouse) 14672, (Rat) 81662
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support