Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEYAGHLPAS SAQGTILACT SCTGAGESGK STIVKQMKII HEDGYSEEEC RQYRAVVYSN TIQSIMAIVK AMGNLQIDFA DPSRADDARQ LFALSCTAEE QGVLPDDLSG VIRRLWADHG VQACFGRSRE YQLNDSAAYY LNDLERIAQS DYIPTQQDVL RTRVKTTGIV ETHFTFKDLH FKMFDVGGQR SERKKWIHCF EGVTAIIFCV ALSAYDLVLA EDEEMNRMHE SMKLFDSICN NKWFTDTSII LFLNKKDLFE EKITHSPLTI CFPEYTGANK YDEAASYIQS KFEDLNKRKD TKEIYTHFTC ATDTKNVQFV FDAVTDVIIK NNLKDCGLF
The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; BR; CD49B; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); GTP-binding protein (G-alpha-i2); GTP-binding regulatory protein Gi alpha-2 chain; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein, alpha inhibiting 2; Guanine nucleotide-binding protein G(i) subunit alpha-2; guanine nucleotide-binding protein G(i), alpha-2 subunit; VLA-2; VLAA2
Gene Aliases: Galphai2; GIP; Gnai-2; GNAI2; GNAI2B; H_LUCA15.1; H_LUCA16.1
UniProt ID: (Human) P04899, (Rat) P04897
Entrez Gene ID: (Human) 2771, (Rat) 81664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support