Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTLESIMAC CLSEEAKEAR RINDEIERQL RRDKRDARRE LKLLLLGTGE SGKSTFIKQM RIIHGSGYSD EDKRGFTKLV YQNIFTAMQA MIRAMDTLKI PYKYEHNKAH AQLVREVDVE KVSAFENPYV DAIKSLWNDP GIQECYDRRR EYQLSDSTKY YLNDLDRVAD PAYLPTQQDV LRVRVPTTGI IEYPFDLQSV IFRMVDVGGQ RSERRKWIHC FENVTSIMFL VALSEYDQVL VESDNENRME ESKALFRTII TYPWFQNSSV ILFLNKKDLL EEKIMYSHLV DYFPEYDGPQ RDAQAAREFI LKMFVDLNPD SDKIIYSHFT CATDTENIRF VFAAVKDTIL QLNLKEYNLV (1-359 aa encoded by BC057777)
Guanine nucleotide-binding proteins are a family of heterotrimeric proteins that couple cell surface, 7-transmembrane domain receptors to intracellular signaling pathways. Receptor activation catalyzes the exchange of GTP for GDP bound to the inactive G protein alpha subunit resulting in a conformational change and dissociation of the complex. The G protein alpha and beta-gamma subunits are capable of regulating various cellular effectors. Activation is terminated by a GTPase intrinsic to the G-alpha subunit. G-alpha-q is the alpha subunit of one of the heterotrimeric GTP-binding proteins that mediates stimulation of phospholipase C-beta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G alpha q; guanine nucleotide binding protein (G protein), q polypeptide; guanine nucleotide binding protein alpha q subunit; guanine nucleotide binding protein, alpha q polypeptide; guanine nucleotide regulatory protein G alpha q; Guanine nucleotide-binding protein alpha-q; Guanine nucleotide-binding protein G(q) subunit alpha; heterotrimeric guanine nucleotide-binding protein alpha q subunit
Gene Aliases: 1110005L02Rik; 6230401I02Rik; AA408290; AW060788; CMC1; Dsk1; Dsk10; G-ALPHA-q; Galphaq; GAQ; GNAQ; Gq; GqI; SWS
UniProt ID: (Human) P50148, (Rat) P82471, (Mouse) P21279
Entrez Gene ID: (Human) 2776, (Rat) 81666, (Mouse) 14682
Molecular Function:
heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support