Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSGSSSVAAM KKVVQQLRLE AGLNRVKVSQ AAADLKQFCL QNAQHDPLLT GVSSSTNPFR PQKVCSFL
G proteins are trimeric (alpha-beta-gamma) membrane-associated proteins that regulate flow of information from cell surface receptors to a variety of internal metabolic effectors. Interaction of a G protein with its activated receptor promotes exchange of GTP for GDP that is bound to the alpha subunit. The alpha-GTP complex dissociates from the beta-gamma heterodimer so that the subunits, in turn, may interact with and regulate effector molecules (Gilman, 1987 [PubMed 3113327];Summary by Ahmad et al., 1995) [PubMed 7606925].[supplied by OMIM, Nov 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: guanine nucleotide binding protein (G protein), gamma 5; Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-5
Gene Aliases: GNG5; GNGT5
UniProt ID: (Human) P63218
Entrez Gene ID: (Human) 2787
Molecular Function:
heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support