Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKPDETPMFD PSLLKEVDWS QNTATFSPAI SPTHPGEGLV LRPLCTADLN RGFFKVLGQL TETGVVSPEQ FMKSFEHMKK SGDYYVTVVE
Glucosamine-6 phosphate-N-acetyl Transferase (GNPNAT1) is a key enzyme in the hexosamine biosynthetic pathway. It converts D-glucosamine 6-phosphate to N-acetyl-D-glucosamine 6-phosphate, the acetylation step allowing carbon and nitrogen enter into the hexosamine biosynthetic pathway. Recently inhibition of GNPNAT1 has been reported to promote proliferation and aggressiveness of castration-resistant prostate cancer. (27194471).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glucosamine 6-phosphate N-acetyltransferase; Phosphoglucosamine acetylase; Phosphoglucosamine transacetylase
Gene Aliases: GNA1; GNPNAT; GNPNAT1; Gpnat1
UniProt ID: (Human) Q96EK6
Entrez Gene ID: (Human) 64841
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support