Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MRPQQAPVSG KVFIQRDYSS GTRCQFQTKF PAELENRIDR QQFEETVRTL NNLYAEAEKL GGQSYLEGCL ACLTAYTIFL CMETHYEKVL KKVSKYIQEQ NEKIYAPQGL LLTDPIERGL RVIEITIYED RGMSSGR
GOLGA7 may be involved in protein transport from Golgi to cell surface. The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: golgi autoantigen, golgin subfamily a, 7; Golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; Golgin subfamily A member 7
Gene Aliases: GCP16; GOLGA3AP1; GOLGA7; GOLGA7A; HDCKB03P; HSPC041
UniProt ID: (Human) Q7Z5G4
Entrez Gene ID: (Human) 51125
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support