Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGLGVSAEQP AGGAEGFHLH GVQENSPAQQ AGLEPYFDFI ITIGHSRLNK ENDTLKALLK ANVEKPVKLE VFNMKTMRVR EVEVVPSNMW GGQGLLGASV RFCSFRRASE QVWHVLDVEP SSPAALAGLR PYTDYVVGSD QILQESEDFF TLIESHEGKP LKLMVYNSKS DSCREVTVTP NAAWGGEGSL GCGIGYGYLH RIPTQPPSYH KKPPGTPPPS ALPLGAPPPD ALPPGPTPED SPSLETGSRQ SDYMEALLQA PGSSMEDPLP GPGSPSHSAP DPDGLPHFME TPLQPPPPVQ RVMDPGFLDV SGISLLDNSN ASVWPSLPSS TELTTTAVST SGPEDICSSS SSHERGGEAT WSGSEFEVSF LDSPGAQAQA DHLPQLTLPD SLTSAASPED GLSAELLEAQ AEEEPASTEG LDTGTEAEGL DSQAQISTTE
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; GRASP65
Gene Aliases: GOLPH5; GORASP1; GRASP65; P65
UniProt ID: (Human) Q9BQQ3
Entrez Gene ID: (Human) 64689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support