Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Glycoprotein-340 is member of the scavenger receptor superfamily and has been identified as a putative SP-D receptor. Gp-340 has been shown to bind SP-D in a calcium dependent manner through a protein-protein interaction. Like SP-D, gp-340 is mainly found in lung tissue and immunohistochemistry revealed high expression in alveolar macrophages. An identical protein is present in saliva (salivary agglutinin) and both forms are encoded by the gene DMBT1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apactin; CRP-ductin; Deleted in malignant brain tumors 1 protein; deleted in malignant brain tumors 1 pseudogene; Ebnerin; Glycoprotein 300; Glycoprotein 340; Gp-340; gp300; Hensin; Mucin-like glycoprotein; Muclin; p80; Pancrin; RP11-481L19.1; SAG; Salivary agglutinin; Surfactant pulmonary-associated D-binding protein; Vomeroglandin
Gene Aliases: CRP; CRP-[a]; CRP-[b]; Crpd; DBMT1; DMBT1; gp300; GP340; muclin; p80; SAG
UniProt ID: (Human) Q9UGM3, (Rat) Q8CIZ5, (Mouse) Q60997
Entrez Gene ID: (Human) 1755, (Rat) 170568, (Mouse) 12945
Molecular Function:
transmembrane signal receptor
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support