Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-21530 detects GPBB in Human samples.
The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycogen phosphorylase B; Glycogen phosphorylase, brain form; MGC9213
Gene Aliases: GPBB; PYGB
UniProt ID: (Human) P11216
Entrez Gene ID: (Human) 5834
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support