Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FFAR; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; G-protein coupled receptor 40; HGNC:4498
Gene Aliases: FFA1R; FFAR1; GPCR40; GPR40
UniProt ID: (Human) O14842, (Rat) Q8K3T4, (Mouse) Q76JU9
Entrez Gene ID: (Human) 2864, (Rat) 266607, (Mouse) 233081
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support