Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is an integral membrane protein and G protein-coupled receptor. The encoded protein is similar in sequence to another G protein-coupled receptor (GPR7), and it is structurally similar to opioid and somatostatin receptors. This protein binds neuropeptides B and W. This gene is intronless and is expressed primarily in the frontal cortex of the brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 8; G-protein coupled receptor 8; Neuropeptides B/W receptor type 2; opioid-somatostatin-like receptor 8
Gene Aliases: GPR8; NPBWR2
UniProt ID: (Human) P48146
Entrez Gene ID: (Human) 2832
Molecular Function: G-protein coupled receptor transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support