Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAFQKAVKGT LVGGGALATV LGLSQFAHYR RKQMNLAYVK AADCISEPVN REPPSREAQL LTLQNTSEFD ILVIGGGATG SGCALDAVTR GLKTALVERD DFSSGTSSRS TKLIHGGVRY LQKAIMKLDI EQYRMVKEAL HERANLLEIA PHLSAPLPIM LPVYKWWQLP YYWVGIKLYD LVAGSNCLKS SYVLSKSRAL EHFPMLQKDK LVGAIVYYDG QHNDARMNLA IALTAARYGA ATANYMEVVS LLKKTDPQTG KVHVSGARCK DVLTGQEFDV RAKCVINATG PFTDSVRKMD DKDAAAICQP SAGVHIVMPG YYSPESMGLL DPATSDGRVI FFLPWQKMTI AGTTDTPTDV THHPIPSEED INFILNEV
Mitochondrial glycerophosphate dehydrogenase (EC 1. 1. 99. 5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycerol-3-phosphate dehydrogenase 2 (mitochondrial); Glycerol-3-phosphate dehydrogenase, mitochondrial; GPD-M; GPDH-M; mGDH; mitochondrial glycerol-3-phosphate dehydrogenase; mitochondrial glycerophosphate dehydrogenase; mitohondrial glycerophosphate dehydrogenase gene; mtGPD; testicular tissue protein Li 76
Gene Aliases: GDH2; GPD2; GPDM; mGPDH
UniProt ID: (Human) P43304
Entrez Gene ID: (Human) 2820
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support