Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VHHLALKDD VRHKVHLNTF GFFKDGYMVV NVSSLSLNEP EDKDVTIGFS LDRTKNDGFS SYLDEDVNYC ILKKQSVSVT LLILDISRSE VRVKSPPEAG TQLPKIIFSR DEKVLGQSQE PNVNPASAGN QTQKTQDGGK SKRSTVDSKA MGEKSFSVHN NGGAVSFQFF FNISTDDQEG LYSLYFHKCL GKELPSDKFT FSLDIEITEK NPDSYLSAGE I (41-260 aa encoded by BC110518)
GPR107 also named as KIAA1624 or LUSTR1 is 600 amino acid protein, which belongs to the LU7TM family. GPR107 localizes in membrane and is a multi-pass membrane protein. GPR107 is a promising candidate receptor for neuronostatin, and neuronostatin, interacting with GPR107, may play an important role in the central control of cardiovascular function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lung seven transmembrane receptor 1; Protein GPR107
Gene Aliases: 9930033D15Rik; AI790205; bA138E2.2; C530034M11; GCDRP; GPR107; KIAA1624; LUSTR1; mKIAA1624
UniProt ID: (Human) Q5VW38, (Mouse) Q8BUV8
Entrez Gene ID: (Human) 57720, (Mouse) 277463
Molecular Function:
G-protein coupled receptor
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support