Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The central nervous system neuropeptide FF (NPFF), is believed to play a role in pain modulation and opiate tolerance as well as other physiological functions. Two G protein-coupled receptors, NPFF1 and NPFF2, were isolated from the human central nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ff1; FLJ10751; G protein-coupled receptor 147; G-protein coupled receptor 147; G-protein coupled receptor NPFF1R; neuropeptide FF 1; Neuropeptide FF receptor 1; NPFF-R1; NPFF1 receptor; RFamide-related peptide receptor; RFamide-related peptide receptor OT7T022
Gene Aliases: Gm236; GPR147; NPFF1; NPFF1R1; NPFFR1; OT7T022
UniProt ID: (Human) Q9GZQ6
Entrez Gene ID: (Human) 64106, (Mouse) 237362, (Rat) 64107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support