Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MMWGAGSPLA WLSAGSGNVN VSSVGPAEGP TGPAAPLPSP KAWDVVLCIS GTLVSCENAL VVAIIVGTPA FRAPMFLLVG SLAVADLLAG LGLVLHFAAV FCIGSAEMSL VLVGVLAMAF TASIGSLLAI TVDRYLSLYN ALTYYSETTV TRTYVMLALV WGGALGLGLL PVLAWNCLDG LTTCGVVYPL SKNHLVVLAI AFFMVFGIML QLYAQICRIV CRHAQQIALQ RHLLPASHYV ATRKGIATLA VVLGAFAACW LPFTVYCLLG DAHSPPLYTY LTLLPATYNS MINPIIYAFR NQDVQKVLWA VCCCCSSSKI PFRSRSPSDV
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACCA orphan receptor; adenylate cyclase constitutive activator; G-protein coupled receptor 3; Hacca; OTTHUMP00000043209
Gene Aliases: ACCA; GPR3
UniProt ID: (Human) P46089
Entrez Gene ID: (Human) 2827
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support