Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GPR52 is a member of the G protein-coupled receptor (GPR) family play important roles in signal transduction from the external environment to the inside of the cell.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axor10; G-protein coupled receptor 52; MGC111751; probable G-protein coupled receptor 52
Gene Aliases: EG620246; GPR52
UniProt ID: (Human) Q9Y2T5, (Mouse) P0C5J4
Entrez Gene ID: (Human) 9293, (Mouse) 620246, (Rat) 684623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support