Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C5AR2 is an orphan member that belongs to anaphylatoxin receptor family. It is a seven transmembrane G-protein-coupled receptor consisting of an N-terminal N-glycosylation site, several conserved cysteines and several potential C-terminal phosphorylation sites. It binds to C5a and C5a-des-Arg and serves as a highly regulated scavenger receptor with a negative modulatory influence on C5aR-mediated inflammation. Northern Blot analysis suggests it is co- expressed along with C5aR in different tissues and cells including neutrophils/macrophages, mast cells, dendritic cells, brain, lung, heart, kidney, liver, ovary and testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; G-protein coupled receptor 77
Gene Aliases: C5AR2; C5L2; GPF77; GPR77
UniProt ID: (Human) Q9P296
Entrez Gene ID: (Human) 27202
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support