Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: AAQALDQYKL RQASIHSNHV ARTDEAMPGR FQELDSRLAS GGPSEGISSE PVSAATTQTL EGDSSEVGDQ INSKRAKQMA EKSPPEASAK AQPIKGARRA PDSSSEFGK
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR EX33 is a member of this family (subfamily Orphan A).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor 84; inflammation-related G protein-coupled receptor EX33; Inflammation-related G-protein coupled receptor EX33
Gene Aliases: EX33; GPCR4; GPR84
UniProt ID: (Human) Q9NQS5
Entrez Gene ID: (Human) 53831
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support