Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitution: 1 X 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
GPR97 is expressed exclusively in pre-B and pre-T cells in the thymus but not in mature lymphoid or non-lymphoid tissues. ESTs have been isolated from spleen libraries. "Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Monkey (94%) Mouse, Rat (89%) Rabbit (83%).".
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor G3; G 97; G protein-coupled receptor 97; G-protein coupled receptor 97; G-protein coupled receptor PGR26; pre B-cell transcription factor 1; probable G-protein coupled receptor 97
Gene Aliases: A030001G24Rik; ADGRG3; AW214383; GPR97; Pb1; PB99; PGR26
UniProt ID: (Human) Q86Y34, (Mouse) Q8R0T6
Entrez Gene ID: (Human) 222487, (Mouse) 54672, (Rat) 291854
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support