Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTGAEIEPS AQAKPEKKAG EEVIAGPERE NDVPLVVRPK VRTQATTGAR PKTETKSVPA ARPKTEAQAM SGARPKTEVQ VMGGARPKTE AQGITGARPK TDARAVGGAR SKTDAKAIPG ARPKDEAQAW AQSEFGTEAV SQAEGVSQTN AVAWPLATAE SGSVTKSKGL SMDRELVNVD AETFPGTQGQ KGIQPWFGPG EETNMGSWCY SRPRAREEAS NESGFWSADE TSTASSFWTG EETSVRSWPR EESNTRSRHR AKHQTNPRSR PRSKQEAYVD SWSGSEDEAS NPFSFWVGEN TNNLFRPRVR EEANIRSKLR TNREDCFESE SEDEFYKQSW VLPGEEANSR FRH (1-352 aa encoded by BC013576)
The protein encoded by this gene is a member of a family that regulates the activity of G protein-coupled receptors (GPCRs). The encoded protein has been shown to be capable of interacting with several GPCRs, including the M1 muscarinic acetylcholine receptor and the calcitonin receptor. Several transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ35662; FLJ37327; G protein-coupled receptor associated sorting protein 2; G-protein coupled receptor-associated sorting protein 2; GASP-2; prion protein ligand 5
Gene Aliases: 5330440H13Rik; GASP-2; GASP2; GPRASP2; Prpl5; RGD1561019
UniProt ID: (Human) Q96D09, (Mouse) Q8BUY8
Entrez Gene ID: (Human) 114928, (Mouse) 245607, (Rat) 317405
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support