Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: ELCILYRSCR QECPLQGNAC PVTAYQHSFQ VENQELSRAR DSDGAEEDVA LTSYGTPIQP QTVDPTQECF IPQAKLSPQQ DAGGV
The protein encoded by this gene is a member of the G protein-coupled receptor family; however, the specific function of this gene has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor, class C, group 5, member D; G protein-coupled receptor, family C, group 5, member D; G-protein coupled receptor family C group 5 member D; MGC129713; MGC129714; orphan G-protein coupled receptor
Gene Aliases: GPRC5D
UniProt ID: (Human) Q9NZD1
Entrez Gene ID: (Human) 55507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support