Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MRDSSAPSSA SSSVTDLYCT PHSSRSDLVL PGTAGDFSLS ASLSACTLLY EGAVEPMQID VDPQEDPQNA PDVNYVVENP SLDLEQYAAS YSGLMRIERL QFIADHCPTL RVEALKMALS FVQRTFNVDM YEEIHRKLSE ATRELQNAPD AIPESGVEPP ALDTAWVEAT RKKALLKLEK LDTDLKNYKG NSIKESIRRG HDDLGDHYLD CGDLSNALKC YSRARDYCTS AKHVINMCLN VIKVSVYLQN WSHVLSYVSK AESTPEIAEQ RGERDSQTQA ILTKLKCAAG LAELAARKYK QAAKCLLLAS FDHCDFPELL SPSNVAIYGG LCALATFDRQ ELQRNVISSS SFKLFLELEP QVRDIIFKFY ESKYASCLKM LDEMKDNLLL DMYLAPHVRT LYTQIRNRAL IQYFSPYVSA DMHRMAAAFN TTVAALEDEL TQLILEGLIS ARVDSHSKIL YARDVDQRST TFEKSLLMGK EFQRRAKAMM LRAAVLRNQI HVKSPPREGS QGELTPANSQ SRMSTNM
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COP9 signalosome complex subunit 1; G protein pathway suppressor 1; GPS-1; JAB1-containing signalosome subunit 1; MGC71287; Protein MFH; SGN1
Gene Aliases: COPS1; CSN1; GPS1; SGN1
UniProt ID: (Human) Q13098
Entrez Gene ID: (Human) 2873
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support