Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MEAERPQEEE DGEQGPPQDE EGWPPPNSTT RPWRSAPPSP PPPGTRHTAL GPRSASLLSL QTELLLDLVA EAQSRRLEEQ RATFYTPQNP SSLAPAPLRP LEDREQLYST ILSHQCQRME AQRSEPPLPP GGQELLELLL RVQGGGRMEE QRSRPPTHTC
Interacts with subunit of G(i) alpha proteins and regulates the activation of G(i) alpha proteins. Target protein is a member of a family of proteins able to activate G-protein signalling in the absence of a receptor; human homolog is located within the MHC class III region; may play a role in immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator of G-protein signaling 4; G-protein signaling modulator 3 (AGS3-like, C. elegans); G-protein signalling modulator 3 (AGS3-like, C. elegans); G-protein-signaling modulator 3; G18.1b; Protein G18
Gene Aliases: AGS4; C6orf9; G18; G18.1a; G18.1b; G18.2; GPSM3; NG1
UniProt ID: (Human) Q9Y4H4
Entrez Gene ID: (Human) 63940
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support