Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GIPADPNNVF LSTGASDAIV TVLKLLVAGE GHTRTGVLIP IPQYPLYSAT LAELGAVQVD YYLDEERAWA LDVAELHRAL GQARDHCRPR ALCVINPGNP TGQVQTRECI EAVIRFAFEE RLFLLADEVY QDNVYAAGSQ FHSFKKVLME MGPPYAGQQE LASFHSTSKG YMGECGFRGG YVEVVNMDAA VQQQMLKLMS VRLCPPVPGQ ALLDLVVSPP APTDPSFAQF QAEKQAVLAE LAAKAKLTEQ VFNEAPGISC NPVQGAMYSF PRVQLPPRAV ERAQELGLAP DMFFCLRLLE ETGICVVPGS GFGQREGTYH FRMTILPPLE KLRLLLEKLS RFHAKFTLEY S
GPT(Glutamate-pyruvate transaminase), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. This protein has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2). See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alanine aminotransferase; Alanine aminotransferase 1; ALT1; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1; Glutamic--pyruvic transaminase 1; glutamic-alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); GPT 1
Gene Aliases: AAT1; ALT1; GPT; GPT1
UniProt ID: (Human) P24298
Entrez Gene ID: (Human) 2875, (Pig) 100524618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support