Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione. May constitute a glutathione peroxidase-like protective system against peroxide damage in sperm membrane lipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arMEP24; EGLP; epididymal androgen-related protein; Epididymal secretory glutathione peroxidase; epididymis secretory sperm binding protein Li 75p; Epididymis-specific glutathione peroxidase-like protein; Glutathione peroxidase 5; GPx-5; GSHPx-5; lysosomal trafficking regulator, long splice form; Major androgen-regulated protein; OTTHUMP00000017919; OTTHUMP00000214636
Gene Aliases: Arep; AV379049; GPX5; HEL-S-75p
UniProt ID: (Human) O75715, (Mouse) P21765, (Rat) P30710
Entrez Gene ID: (Human) 2880, (Mouse) 14780, (Rat) 113919
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support