Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MGLGVSAEQ PAGGAEGFHL HGVQENSPAQ QAGLEPYFDF IITIGHSRLN KENDTLKALL KANVEKPVKL EVFNMKTMRV REVEVVPSNM WGGQGLLGAS VRFCSFRRAS EQVWHVLDVE PSSPAALAGL RPYTDYVVGS DQILQESEDF FTLIESHEGK PLKLMVYNSK SDSCRESGMW HWLWVSTPDP NSAPQLPQEA TWHPTTFCST TWCPTT (1-215 aa encoded by B C008928)
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; GRASP65
Gene Aliases: 5430411C10Rik; GOLPH5; GORASP1; GRASP65; P65
UniProt ID: (Human) Q9BQQ3, (Mouse) Q91X51
Entrez Gene ID: (Human) 64689, (Rat) 56082, (Mouse) 74498
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support