Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MFWKLSLSLF LVAVLVKVAE ARKNRPAGAI PSPYKDGSSN NSERWQHQIK
Sequence homology: Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 93%
GREM2 encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CKTSF1B2FLJ21195; Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; DAND3Prdc; gremlin 2, cysteine knot superfamily, homolog; Gremlin-2; PRDCgremlin 2; Protein related to DAN and cerberus
Gene Aliases: CKTSF1B2; DAND3; GREM2; PRDC
UniProt ID: (Human) Q9H772
Entrez Gene ID: (Human) 64388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support