Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycerate-2-dehydrogenase; Glyoxylate reductase/hydroxypyruvate reductase
Gene Aliases: GLXR; GLYD; GRHPR; MSTP035; PH2
UniProt ID: (Human) Q9UBQ7
Entrez Gene ID: (Human) 9380
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support