Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VRLDEREFPE AKNAPLKYTS ALTHDAILVI AEAFRYLRRQ RVDVSRRGSA GDCLANPAVP WSQGIDIERA LKMVQVQGMT GNIQFDTYGR RTNYTIDVYE MKVSGSRKAG YWNEYERFVP FSDQQISNDS ASSENRTI
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMPA-selective glutamate receptor 3; dJ1171F9.1; GluR-3; GluR-C; GluR-K3; glutamate receptor; Glutamate receptor 3; glutamate receptor C; glutamate receptor channel alpha3 subunit; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA3 (alpha 3); OTTHUMP00000024262; RP11-349N19.3
Gene Aliases: 2900064I19Rik; GluA3; GluR-3; GLUR-C; GLUR-K3; GLUR3; Gluralpha3; GLURC; GRIA3; Kiaa4184; MRX94
UniProt ID: (Human) P42263, (Mouse) Q9Z2W9, (Rat) P19492
Entrez Gene ID: (Human) 2892, (Mouse) 53623, (Rat) 29628
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support