Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAASKVKQD MPPPGGYGPI DYKRNLPRRG LSGYSMLAIG IGTLIYGHWS IMKWNRERRR LQIEDFEARI ALLPLLQAET DRRTLQMLRE NLEEEAIIMK DVPDWKVGES VFHTTRWVPP LIGELYGLRT TEEALHASHG FMWYT (1-144 aa encoded by BC009189)
A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CI-B16.6; complex I B16.6 subunit; Complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; Gene associated with retinoic and interferon-induced mortality 19 protein; gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM-19; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH-ubiquinone oxidoreductase B16.6 subunit
Gene Aliases: 2700054G14Rik; AU022060; B16.6; CDA016; CGI-39; GRIM-19; GRIM19; NDUFA13
UniProt ID: (Human) Q9P0J0, (Mouse) Q9ERS2
Entrez Gene ID: (Rat) 100911483, (Human) 51079, (Mouse) 67184
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support