Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AVELLDGQS KTKGYAGTPG FMAPELLQGE EYDFSVDYFA LGVTLYEMIA ARGPFRARGE KVENKELKHR IISEPVKYPD KFSQASKDFC EALLEKDPEK RLGFRDETCD KLRAHPLFKD LNWRQLEAGM LMPPFIPDSK TVYAKDIQDV GAFSTVKGVA FDKTDTEFFQ EFATGNCPIP WQEEMIETGI FGELNVWRSD GQMPDDMKGI SGGSSSSSKS GMCLVS (339-563 aa encoded by BC140945)
GRK1 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinases. GRK1 mediates rapid desensitization of rod photoreceptors to light by catalyzing phosphorylation of the visual pigment rhodopsin which leads to its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). GRK1 can be phosphorylated by PKA and this reduces the ability of GRK1 to phosphorylate rhodopsin in vitro. It is speculated that phosphorylation of GRK1 by PKA in vivo occurs in the dark when cAMP levels in photoreceptor cells are elevated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor kinase 1; G-protein receptor kinase 1; GRK 1a/b; GRK1a; rhodopsin kinase; Rhodopsin kinase GRK1; RK
Gene Aliases: GPRK1; GRK1; RHOK; RK
UniProt ID: (Human) Q15835
Entrez Gene ID: (Human) 6011, (Mouse) 24013
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support