Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: LGYNNYFYGQ LHVQAAPVGP ACCGAVPPLG AQQCSCVPTP PGYEGPGSVL VSPVPHQMLP YMNVGTLSRT ELQLLNQLHC RRKRRHRTIF TDEQLEALEN LFQETKYPD
This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein goosecoid
Gene Aliases: GSC; SAMS
UniProt ID: (Human) P56915
Entrez Gene ID: (Human) 145258
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support