Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive control: PC-3 cell lysate, NIH/3T3 cell lysate, THP-1 cell lysate, THP-1 cell lysate treated with PMA for 17.5 hours and then treated with LPS for 6 hours, PC-12 cell lysate, PMVEC cell lysate, RH-35 cell lysate, L6 cell lysate.
Predicted band size: 53/30 kDa
Subcellular Location: Cell membrane, Secreted.
GSDMD (gasdermin-D) promotes pyroptosis in response to microbial infection and danger signals. It is produced by the cleavage of gasdermin-D by inflammatory caspases CASP1 or CASP4 in response to canonical, as well as, non-canonical (such as cytosolic LPS) inflammasome activators. After cleavage, GSDMD moves to the plasma membrane where it strongly binds to inner leaflet lipids including phosphatidylinositol 4-phosphate, phosphatidylinositol (4,5)-biphosphate, phosphatidylinositol (3,4,5)-biphosphate, phosphatidic acid, and phosphatidylserine. GSDMD also exhibits bactericidal activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gasdermin domain containing 1; Gasdermin domain-containing protein 1; Gasdermin-D
Gene Aliases: 1810036L03Rik; AW558049; DF5L; DFNA5L; FKSG10; GSDMD; GSDMDC1; M2-4
UniProt ID: (Human) P57764, (Mouse) Q9D8T2
Entrez Gene ID: (Human) 79792, (Rat) 315084, (Mouse) 69146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support