Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MFSVFEEITR IVVKEMDAGG DMIAVRSLVD ADRFRCFHLV GEKRTFFGCR HYTTGLTLMD ILDTDGDKWL DELDSGLQGQ KAEFQILDNV DSTGELIVRL PKEITISGSF QGFHHQKIKI SENRISQQYL ATLENRKLKR ELPFSFRSIN TRENLYLVTE TLETVKEETL KSDRQYKFWS QISQGHLSYK HKGQREVTIP PNRVLSYRVK QLVFPNKETM SAGLDIHFRG KTKSFPEGKS LGSEDSRNMK EKLEDMESVL KDLTEEKRKD VLNSLAKCLG KEDIRQDLEQ RVSEVLISRE LHMEDSDKPL LSSLFNAAGV LVEARAKAIL DFLDALLELS EEQQFVAEAL EKGTLPLLKD QVKSVMEQNW DELASSPPDM DYDPEARILC ALYVVVSILL ELAEGPTSVS S
This gene encodes a member of the gasdermin-domain containing protein family. Other gasdermin-family genes are implicated in the regulation of apoptosis in epithelial cells, and are linked to cancer. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but they are candidates for nonsense-mediated mRNA decay (NMD) and are unlikely to be protein-coding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gasdermin-B; Gasdermin-like protein
Gene Aliases: GSDMB; GSDML; PP4052; PRO2521
UniProt ID: (Human) Q8TAX9
Entrez Gene ID: (Human) 55876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support