Product References
Lactobacillus plantarum ZY08 relieves chronic alcohol-induced hepatic steatosis and liver injury in mice via restoring intestinal flora homeostasis.
Food research international (Ottawa, Ont.)
Ding Q,Cao F,Lai S,Zhuge H,Chang K,Valencak TG,Liu J,Li S,Ren D
Fri Jul 01 00:00:00 EDT 2022
A postpartum enriched environment rescues impaired cognition and oxidative markers in aged mice with gestational inflammation.
Brain and behavior
Zhang YX,Wei QY,Wang YT,Zeng LP,Sun SY,Wu YF,Ren CY,Wang F,Chen GH,Cao L
Thu Dec 01 00:00:00 EST 2022
S-Nitrosoglutathione Reductase Deficiency Causes Aberrant Placental S-Nitrosylation and Preeclampsia.
Journal of the American Heart Association
Kulandavelu S,Dulce RA,Murray CI,Bellio MA,Fritsch J,Kanashiro-Takeuchi R,Arora H,Paulino E,Soetkamp D,Balkan W,Van Eyk JE,Hare JM
Tue Mar 01 00:00:00 EST 2022
Analysis of disease model iPSCs derived from patients with a novel Fanconi anemia-like IBMFS ADH5/ALDH2 deficiency.
Blood
Mu A,Hira A,Niwa A,Osawa M,Yoshida K,Mori M,Okamoto Y,Inoue K,Kondo K,Kanemaki MT,Matsuda T,Ito E,Kojima S,Nakahata T,Ogawa S,Tanaka K,Matsuo K,Saito MK,Takata M
Thu Apr 15 00:00:00 EDT 2021
S-nitrosoglutathione reductase maintains mitochondrial homeostasis by promoting clearance of damaged mitochondria in porcine preimplantation embryos.
Cell proliferation
Niu YJ,Zhou D,Cui XS
Mon Mar 01 00:00:00 EST 2021
S-nitrosoglutathione reductase maintains mitochondrial homeostasis by promoting clearance of damaged mitochondria in porcine preimplantation embryos.
Cell proliferation
Niu YJ,Zhou D,Cui XS
Mon Mar 01 00:00:00 EST 2021
S-nitrosoglutathione reductase (GSNOR) deficiency accelerates cardiomyocyte differentiation of induced pluripotent stem cells.
The journal of cardiovascular aging
Salerno AG,Wanschel ACBA,Dulce RA,Hatzistergos KE,Balkan W,Hare JM
Thu Apr 28 00:00:00 EDT 2022
Bronchopulmonary Dysplasia and Pulmonary Hypertension. The Role of Smooth Muscle adh5.
American journal of respiratory cell and molecular biology
Raffay TM,Bonilla-Fernandez K,Jafri A,Sopi RB,Smith LA,Cui F,O'Reilly M,Zhang R,Hodges CA,MacFarlane PM,Deutsch G,Martin RJ,Gaston B
Thu Jul 01 00:00:00 EDT 2021
Two Aldehyde Clearance Systems Are Essential to Prevent Lethal Formaldehyde Accumulation in Mice and Humans.
Molecular cell
Dingler FA,Wang M,Mu A,Millington CL,Oberbeck N,Watcham S,Pontel LB,Kamimae-Lanning AN,Langevin F,Nadler C,Cordell RL,Monks PS,Yu R,Wilson NK,Hira A,Yoshida K,Mori M,Okamoto Y,Okuno Y,Muramatsu H,Shiraishi Y,Kobayashi M,Moriguchi T,Osumi T,Kato M,Miyano S,Ito E,Kojima S,Yabe H,Yabe M,Matsuo K,Ogawa S,Göttgens B,Hodskinson MRG,Takata M,Patel KJ
Thu Dec 17 00:00:00 EST 2020
Two Aldehyde Clearance Systems Are Essential to Prevent Lethal Formaldehyde Accumulation in Mice and Humans.
Molecular cell
Dingler FA,Wang M,Mu A,Millington CL,Oberbeck N,Watcham S,Pontel LB,Kamimae-Lanning AN,Langevin F,Nadler C,Cordell RL,Monks PS,Yu R,Wilson NK,Hira A,Yoshida K,Mori M,Okamoto Y,Okuno Y,Muramatsu H,Shiraishi Y,Kobayashi M,Moriguchi T,Osumi T,Kato M,Miyano S,Ito E,Kojima S,Yabe H,Yabe M,Matsuo K,Ogawa S,Göttgens B,Hodskinson MRG,Takata M,Patel KJ
Thu Dec 17 00:00:00 EST 2020
Two Aldehyde Clearance Systems Are Essential to Prevent Lethal Formaldehyde Accumulation in Mice and Humans.
Molecular cell
Dingler FA,Wang M,Mu A,Millington CL,Oberbeck N,Watcham S,Pontel LB,Kamimae-Lanning AN,Langevin F,Nadler C,Cordell RL,Monks PS,Yu R,Wilson NK,Hira A,Yoshida K,Mori M,Okamoto Y,Okuno Y,Muramatsu H,Shiraishi Y,Kobayashi M,Moriguchi T,Osumi T,Kato M,Miyano S,Ito E,Kojima S,Yabe H,Yabe M,Matsuo K,Ogawa S,Göttgens B,Hodskinson MRG,Takata M,Patel KJ
Thu Dec 17 00:00:00 EST 2020
Two Aldehyde Clearance Systems Are Essential to Prevent Lethal Formaldehyde Accumulation in Mice and Humans.
Molecular cell
Dingler FA,Wang M,Mu A,Millington CL,Oberbeck N,Watcham S,Pontel LB,Kamimae-Lanning AN,Langevin F,Nadler C,Cordell RL,Monks PS,Yu R,Wilson NK,Hira A,Yoshida K,Mori M,Okamoto Y,Okuno Y,Muramatsu H,Shiraishi Y,Kobayashi M,Moriguchi T,Osumi T,Kato M,Miyano S,Ito E,Kojima S,Yabe H,Yabe M,Matsuo K,Ogawa S,Göttgens B,Hodskinson MRG,Takata M,Patel KJ
Thu Dec 17 00:00:00 EST 2020
Cyclic compression increases F508 Del CFTR expression in ciliated human airway epithelium.
American journal of physiology. Lung cellular and molecular physiology
Marozkina N,Bosch J,Cotton C,Smith L,Seckler J,Zaman K,Rehman S,Periasamy A,Gaston H,Altawallbeh G,Davis M,Jones DR,Schilz R,Randell SH,Gaston B
Thu Aug 01 00:00:00 EDT 2019
S-nitrosylation of endothelial nitric oxide synthase impacts erectile function.
International journal of impotence research
Kavoussi PK,Smith RP,Oliver JL,Costabile RA,Steers WD,Brown-Steinke K,de Ronde K,Lysiak JJ,Palmer LA
Tue Jan 01 00:00:00 EST 2019
S-nitrosylation of endothelial nitric oxide synthase impacts erectile function.
International journal of impotence research
Kavoussi PK,Smith RP,Oliver JL,Costabile RA,Steers WD,Brown-Steinke K,de Ronde K,Lysiak JJ,Palmer LA
Tue Jan 01 00:00:00 EST 2019
S-nitrosylation of endothelial nitric oxide synthase impacts erectile function.
International journal of impotence research
Kavoussi PK,Smith RP,Oliver JL,Costabile RA,Steers WD,Brown-Steinke K,de Ronde K,Lysiak JJ,Palmer LA
Tue Jan 01 00:00:00 EST 2019
S-nitrosylation of endothelial nitric oxide synthase impacts erectile function.
International journal of impotence research
Kavoussi PK,Smith RP,Oliver JL,Costabile RA,Steers WD,Brown-Steinke K,de Ronde K,Lysiak JJ,Palmer LA
Tue Jan 01 00:00:00 EST 2019
AKR1A1 is a novel mammalian S-nitroso-glutathione reductase.
The Journal of biological chemistry
Stomberski CT,Anand P,Venetos NM,Hausladen A,Zhou HL,Premont RT,Stamler JS
Fri Nov 29 00:00:00 EST 2019
Phenylpyrrole fungicides act on triosephosphate isomerase to induce methylglyoxal stress and alter hybrid histidine kinase activity.
Scientific reports
Brandhorst TT,Kean IRL,Lawry SM,Wiesner DL,Klein BS
Mon Mar 25 00:00:00 EDT 2019
GSNOR modulates hyperhomocysteinemia-induced T cell activation and atherosclerosis by switching Akt S-nitrosylation to phosphorylation.
Redox biology
Li J,Zhang Y,Zhang Y,Lü S,Miao Y,Yang J,Huang S,Ma X,Han L,Deng J,Fan F,Liu B,Huo Y,Xu Q,Chen C,Wang X,Feng J
Sun Jul 01 00:00:00 EDT 2018
GSNOR modulates hyperhomocysteinemia-induced T cell activation and atherosclerosis by switching Akt S-nitrosylation to phosphorylation.
Redox biology
Li J,Zhang Y,Zhang Y,Lü S,Miao Y,Yang J,Huang S,Ma X,Han L,Deng J,Fan F,Liu B,Huo Y,Xu Q,Chen C,Wang X,Feng J
Sun Jul 01 00:00:00 EDT 2018
GSNOR Deficiency Enhances In Situ Skeletal Muscle Strength, Fatigue Resistance, and RyR1 S-Nitrosylation Without Impacting Mitochondrial Content and Activity.
Antioxidants & redox signaling
Moon Y,Cao Y,Zhu J,Xu Y,Balkan W,Buys ES,Diaz F,Kerrick WG,Hare JM,Percival JM
Wed Feb 01 00:00:00 EST 2017
GSNOR Deficiency Enhances In Situ Skeletal Muscle Strength, Fatigue Resistance, and RyR1 S-Nitrosylation Without Impacting Mitochondrial Content and Activity.
Antioxidants & redox signaling
Moon Y,Cao Y,Zhu J,Xu Y,Balkan W,Buys ES,Diaz F,Kerrick WG,Hare JM,Percival JM
Wed Feb 01 00:00:00 EST 2017
Balance between S-nitrosylation and denitrosylation modulates myoblast proliferation independently of soluble guanylyl cyclase activation.
American journal of physiology. Cell physiology
Yamashita AMS,Ancillotti MTC,Rangel LP,Fontenele M,Figueiredo-Freitas C,Possidonio AC,Soares CP,Sorenson MM,Mermelstein C,Nogueira L
Sat Jul 01 00:00:00 EDT 2017
Balance between S-nitrosylation and denitrosylation modulates myoblast proliferation independently of soluble guanylyl cyclase activation.
American journal of physiology. Cell physiology
Yamashita AMS,Ancillotti MTC,Rangel LP,Fontenele M,Figueiredo-Freitas C,Possidonio AC,Soares CP,Sorenson MM,Mermelstein C,Nogueira L
Sat Jul 01 00:00:00 EDT 2017
Balance between S-nitrosylation and denitrosylation modulates myoblast proliferation independently of soluble guanylyl cyclase activation.
American journal of physiology. Cell physiology
Yamashita AMS,Ancillotti MTC,Rangel LP,Fontenele M,Figueiredo-Freitas C,Possidonio AC,Soares CP,Sorenson MM,Mermelstein C,Nogueira L
Sat Jul 01 00:00:00 EDT 2017
Balance between S-nitrosylation and denitrosylation modulates myoblast proliferation independently of soluble guanylyl cyclase activation.
American journal of physiology. Cell physiology
Yamashita AMS,Ancillotti MTC,Rangel LP,Fontenele M,Figueiredo-Freitas C,Possidonio AC,Soares CP,Sorenson MM,Mermelstein C,Nogueira L
Sat Jul 01 00:00:00 EDT 2017
Alcohol Dehydrogenase 5 Is a Source of Formate for De Novo Purine Biosynthesis in HepG2 Cells.
The Journal of nutrition
Bae S,Chon J,Field MS,Stover PJ
Sat Apr 01 00:00:00 EDT 2017
Alcohol Dehydrogenase 5 Is a Source of Formate for De Novo Purine Biosynthesis in HepG2 Cells.
The Journal of nutrition
Bae S,Chon J,Field MS,Stover PJ
Sat Apr 01 00:00:00 EDT 2017
S-nitrosylation of the Mitochondrial Chaperone TRAP1 Sensitizes Hepatocellular Carcinoma Cells to Inhibitors of Succinate Dehydrogenase.
Cancer research
Rizza S,Montagna C,Cardaci S,Maiani E,Di Giacomo G,Sanchez-Quiles V,Blagoev B,Rasola A,De Zio D,Stamler JS,Cecconi F,Filomeni G
Fri Jul 15 00:00:00 EDT 2016
S-nitrosylation of the Mitochondrial Chaperone TRAP1 Sensitizes Hepatocellular Carcinoma Cells to Inhibitors of Succinate Dehydrogenase.
Cancer research
Rizza S,Montagna C,Cardaci S,Maiani E,Di Giacomo G,Sanchez-Quiles V,Blagoev B,Rasola A,De Zio D,Stamler JS,Cecconi F,Filomeni G
Fri Jul 15 00:00:00 EDT 2016
De Novo-Synthesized Retinoic Acid in Ovarian Antral Follicles Enhances FSH-Mediated Ovarian Follicular Cell Differentiation and Female Fertility.
Endocrinology
Kawai T,Yanaka N,Richards JS,Shimada M
Sun May 01 00:00:00 EDT 2016
De Novo-Synthesized Retinoic Acid in Ovarian Antral Follicles Enhances FSH-Mediated Ovarian Follicular Cell Differentiation and Female Fertility.
Endocrinology
Kawai T,Yanaka N,Richards JS,Shimada M
Sun May 01 00:00:00 EDT 2016
Copper disrupts S-nitrosothiol signaling in activated BV2 microglia.
Neurochemistry international
Rossi-George A,Guo CJ
Sat Oct 01 00:00:00 EDT 2016
Copper disrupts S-nitrosothiol signaling in activated BV2 microglia.
Neurochemistry international
Rossi-George A,Guo CJ
Sat Oct 01 00:00:00 EDT 2016
S-Nitrosoglutathione Attenuates Airway Hyperresponsiveness in Murine Bronchopulmonary Dysplasia.
Molecular pharmacology
Raffay TM,Dylag AM,Di Fiore JM,Smith LA,Einisman HJ,Li Y,Lakner MM,Khalil AM,MacFarlane PM,Martin RJ,Gaston B
Sat Oct 01 00:00:00 EDT 2016
S-Nitrosoglutathione Attenuates Airway Hyperresponsiveness in Murine Bronchopulmonary Dysplasia.
Molecular pharmacology
Raffay TM,Dylag AM,Di Fiore JM,Smith LA,Einisman HJ,Li Y,Lakner MM,Khalil AM,MacFarlane PM,Martin RJ,Gaston B
Sat Oct 01 00:00:00 EDT 2016
Augmentation of CFTR maturation by S-nitrosoglutathione reductase.
American journal of physiology. Lung cellular and molecular physiology
Zaman K,Sawczak V,Zaidi A,Butler M,Bennett D,Getsy P,Zeinomar M,Greenberg Z,Forbes M,Rehman S,Jyothikumar V,DeRonde K,Sattar A,Smith L,Corey D,Straub A,Sun F,Palmer L,Periasamy A,Randell S,Kelley TJ,Lewis SJ,Gaston B
Mon Feb 01 00:00:00 EST 2016
Augmentation of CFTR maturation by S-nitrosoglutathione reductase.
American journal of physiology. Lung cellular and molecular physiology
Zaman K,Sawczak V,Zaidi A,Butler M,Bennett D,Getsy P,Zeinomar M,Greenberg Z,Forbes M,Rehman S,Jyothikumar V,DeRonde K,Sattar A,Smith L,Corey D,Straub A,Sun F,Palmer L,Periasamy A,Randell S,Kelley TJ,Lewis SJ,Gaston B
Mon Feb 01 00:00:00 EST 2016
Phenotype of asthmatics with increased airway S-nitrosoglutathione reductase activity.
The European respiratory journal
Marozkina NV,Wang XQ,Stsiapura V,Fitzpatrick A,Carraro S,Hawkins GA,Bleecker E,Meyers D,Jarjour N,Fain SB,Wenzel S,Busse W,Castro M,Panettieri RA,Moore W,Lewis SJ,Palmer LA,Altes T,de Lange EE,Erzurum S,Teague WG,Gaston B
Thu Jan 01 00:00:00 EST 2015
Phenotype of asthmatics with increased airway S-nitrosoglutathione reductase activity.
The European respiratory journal
Marozkina NV,Wang XQ,Stsiapura V,Fitzpatrick A,Carraro S,Hawkins GA,Bleecker E,Meyers D,Jarjour N,Fain SB,Wenzel S,Busse W,Castro M,Panettieri RA,Moore W,Lewis SJ,Palmer LA,Altes T,de Lange EE,Erzurum S,Teague WG,Gaston B
Thu Jan 01 00:00:00 EST 2015
Phenotype of asthmatics with increased airway S-nitrosoglutathione reductase activity.
The European respiratory journal
Marozkina NV,Wang XQ,Stsiapura V,Fitzpatrick A,Carraro S,Hawkins GA,Bleecker E,Meyers D,Jarjour N,Fain SB,Wenzel S,Busse W,Castro M,Panettieri RA,Moore W,Lewis SJ,Palmer LA,Altes T,de Lange EE,Erzurum S,Teague WG,Gaston B
Thu Jan 01 00:00:00 EST 2015
Phenotype of asthmatics with increased airway S-nitrosoglutathione reductase activity.
The European respiratory journal
Marozkina NV,Wang XQ,Stsiapura V,Fitzpatrick A,Carraro S,Hawkins GA,Bleecker E,Meyers D,Jarjour N,Fain SB,Wenzel S,Busse W,Castro M,Panettieri RA,Moore W,Lewis SJ,Palmer LA,Altes T,de Lange EE,Erzurum S,Teague WG,Gaston B
Thu Jan 01 00:00:00 EST 2015
Hypoxia-induced changes in protein s-nitrosylation in female mouse brainstem.
American journal of respiratory cell and molecular biology
Palmer LA,Brown-Steinke K,Gunter S,Jyothikumar V,Forbes MS,Lewis SJ
Thu Jan 01 00:00:00 EST 2015
Hypoxia-induced changes in protein s-nitrosylation in female mouse brainstem.
American journal of respiratory cell and molecular biology
Palmer LA,Brown-Steinke K,Gunter S,Jyothikumar V,Forbes MS,Lewis SJ
Thu Jan 01 00:00:00 EST 2015
Pharmacological In Vivo Inhibition of S-Nitrosoglutathione Reductase Attenuates Bleomycin-Induced Inflammation and Fibrosis.
The Journal of pharmacology and experimental therapeutics
Luzina IG,Lockatell V,Todd NW,Kopach P,Pentikis HS,Atamas SP
Thu Oct 01 00:00:00 EDT 2015
Pharmacological In Vivo Inhibition of S-Nitrosoglutathione Reductase Attenuates Bleomycin-Induced Inflammation and Fibrosis.
The Journal of pharmacology and experimental therapeutics
Luzina IG,Lockatell V,Todd NW,Kopach P,Pentikis HS,Atamas SP
Thu Oct 01 00:00:00 EDT 2015
S-nitrosoglutathione inhibits inducible nitric oxide synthase upregulation by redox posttranslational modification in experimental diabetic retinopathy.
Investigative ophthalmology & visual science
Rosales MA,Silva KC,Duarte DA,de Oliveira MG,de Souza GF,Catharino RR,Ferreira MS,Lopes de Faria JB,Lopes de Faria JM
Fri May 02 00:00:00 EDT 2014
S-nitrosoglutathione inhibits inducible nitric oxide synthase upregulation by redox posttranslational modification in experimental diabetic retinopathy.
Investigative ophthalmology & visual science
Rosales MA,Silva KC,Duarte DA,de Oliveira MG,de Souza GF,Catharino RR,Ferreira MS,Lopes de Faria JB,Lopes de Faria JM
Fri May 02 00:00:00 EDT 2014
The role of regulatory proteins and S-nitrosylation of endothelial nitric oxide synthase in the human clitoris: implications for female sexual function.
The journal of sexual medicine
Oliver JL,Kavoussi PK,Smith RP,Woodson RI,Corbett ST,Costabile RA,Palmer LA,Lysiak JJ
Fri Aug 01 00:00:00 EDT 2014
The role of regulatory proteins and S-nitrosylation of endothelial nitric oxide synthase in the human clitoris: implications for female sexual function.
The journal of sexual medicine
Oliver JL,Kavoussi PK,Smith RP,Woodson RI,Corbett ST,Costabile RA,Palmer LA,Lysiak JJ
Fri Aug 01 00:00:00 EDT 2014
The role of regulatory proteins and S-nitrosylation of endothelial nitric oxide synthase in the human clitoris: implications for female sexual function.
The journal of sexual medicine
Oliver JL,Kavoussi PK,Smith RP,Woodson RI,Corbett ST,Costabile RA,Palmer LA,Lysiak JJ
Fri Aug 01 00:00:00 EDT 2014
The role of regulatory proteins and S-nitrosylation of endothelial nitric oxide synthase in the human clitoris: implications for female sexual function.
The journal of sexual medicine
Oliver JL,Kavoussi PK,Smith RP,Woodson RI,Corbett ST,Costabile RA,Palmer LA,Lysiak JJ
Fri Aug 01 00:00:00 EDT 2014
A novel suppressive effect of alcohol dehydrogenase 5 in neuronal differentiation.
The Journal of biological chemistry
Wu K,Ren R,Su W,Wen B,Zhang Y,Yi F,Qiao X,Yuan T,Wang J,Liu L,Izpisua Belmonte JC,Liu GH,Chen C
Fri Jul 18 00:00:00 EDT 2014
A novel suppressive effect of alcohol dehydrogenase 5 in neuronal differentiation.
The Journal of biological chemistry
Wu K,Ren R,Su W,Wen B,Zhang Y,Yi F,Qiao X,Yuan T,Wang J,Liu L,Izpisua Belmonte JC,Liu GH,Chen C
Fri Jul 18 00:00:00 EDT 2014
Treatment with a highly selective β₁ antagonist causes dose-dependent impairment of cerebral perfusion after hemodilution in rats.
Anesthesia and analgesia
Hu T,Beattie WS,Mazer CD,Leong-Poi H,Fujii H,Wilson DF,Tsui AK,Liu E,Muhammad M,Baker AJ,Hare GM
Fri Mar 01 00:00:00 EST 2013
Treatment with a highly selective β₁ antagonist causes dose-dependent impairment of cerebral perfusion after hemodilution in rats.
Anesthesia and analgesia
Hu T,Beattie WS,Mazer CD,Leong-Poi H,Fujii H,Wilson DF,Tsui AK,Liu E,Muhammad M,Baker AJ,Hare GM
Fri Mar 01 00:00:00 EST 2013
Reduction of cardiomyocyte S-nitrosylation by S-nitrosoglutathione reductase protects against sepsis-induced myocardial depression.
American journal of physiology. Heart and circulatory physiology
Sips PY,Irie T,Zou L,Shinozaki S,Sakai M,Shimizu N,Nguyen R,Stamler JS,Chao W,Kaneki M,Ichinose F
Mon Apr 15 00:00:00 EDT 2013
Reduction of cardiomyocyte S-nitrosylation by S-nitrosoglutathione reductase protects against sepsis-induced myocardial depression.
American journal of physiology. Heart and circulatory physiology
Sips PY,Irie T,Zou L,Shinozaki S,Sakai M,Shimizu N,Nguyen R,Stamler JS,Chao W,Kaneki M,Ichinose F
Mon Apr 15 00:00:00 EDT 2013
Reduction of cardiomyocyte S-nitrosylation by S-nitrosoglutathione reductase protects against sepsis-induced myocardial depression.
American journal of physiology. Heart and circulatory physiology
Sips PY,Irie T,Zou L,Shinozaki S,Sakai M,Shimizu N,Nguyen R,Stamler JS,Chao W,Kaneki M,Ichinose F
Mon Apr 15 00:00:00 EDT 2013
Reduction of cardiomyocyte S-nitrosylation by S-nitrosoglutathione reductase protects against sepsis-induced myocardial depression.
American journal of physiology. Heart and circulatory physiology
Sips PY,Irie T,Zou L,Shinozaki S,Sakai M,Shimizu N,Nguyen R,Stamler JS,Chao W,Kaneki M,Ichinose F
Mon Apr 15 00:00:00 EDT 2013
An in vitro model of human acute ethanol exposure that incorporates CXCR3- and CXCR4-dependent recruitment of immune cells.
Toxicological sciences : an official journal of the Society of Toxicology
Karim S,Liaskou E,Hadley S,Youster J,Faint J,Adams DH,Lalor PF
Fri Mar 01 00:00:00 EST 2013
An in vitro model of human acute ethanol exposure that incorporates CXCR3- and CXCR4-dependent recruitment of immune cells.
Toxicological sciences : an official journal of the Society of Toxicology
Karim S,Liaskou E,Hadley S,Youster J,Faint J,Adams DH,Lalor PF
Fri Mar 01 00:00:00 EST 2013
Pharmacological inhibition of S-nitrosoglutathione reductase improves endothelial vasodilatory function in rats in vivo.
Journal of applied physiology (Bethesda, Md. : 1985)
Chen Q,Sievers RE,Varga M,Kharait S,Haddad DJ,Patton AK,Delany CS,Mutka SC,Blonder JP,Dubé GP,Rosenthal GJ,Springer ML
Fri Mar 15 00:00:00 EDT 2013
Pharmacological inhibition of S-nitrosoglutathione reductase improves endothelial vasodilatory function in rats in vivo.
Journal of applied physiology (Bethesda, Md. : 1985)
Chen Q,Sievers RE,Varga M,Kharait S,Haddad DJ,Patton AK,Delany CS,Mutka SC,Blonder JP,Dubé GP,Rosenthal GJ,Springer ML
Fri Mar 15 00:00:00 EDT 2013
S-nitrosoglutathione reductase in human lung cancer.
American journal of respiratory cell and molecular biology
Marozkina NV,Wei C,Yemen S,Wallrabe H,Nagji AS,Liu L,Morozkina T,Jones DR,Gaston B
Sun Jan 01 00:00:00 EST 2012
S-nitrosoglutathione reductase in human lung cancer.
American journal of respiratory cell and molecular biology
Marozkina NV,Wei C,Yemen S,Wallrabe H,Nagji AS,Liu L,Morozkina T,Jones DR,Gaston B
Sun Jan 01 00:00:00 EST 2012
S-nitrosoglutathione reductase in human lung cancer.
American journal of respiratory cell and molecular biology
Marozkina NV,Wei C,Yemen S,Wallrabe H,Nagji AS,Liu L,Morozkina T,Jones DR,Gaston B
Sun Jan 01 00:00:00 EST 2012
S-nitrosoglutathione reductase in human lung cancer.
American journal of respiratory cell and molecular biology
Marozkina NV,Wei C,Yemen S,Wallrabe H,Nagji AS,Liu L,Morozkina T,Jones DR,Gaston B
Sun Jan 01 00:00:00 EST 2012
S-nitrosoglutathione reductase in human lung cancer.
American journal of respiratory cell and molecular biology
Marozkina NV,Wei C,Yemen S,Wallrabe H,Nagji AS,Liu L,Morozkina T,Jones DR,Gaston B
Sun Jan 01 00:00:00 EST 2012
S-nitrosoglutathione reductase in human lung cancer.
American journal of respiratory cell and molecular biology
Marozkina NV,Wei C,Yemen S,Wallrabe H,Nagji AS,Liu L,Morozkina T,Jones DR,Gaston B
Sun Jan 01 00:00:00 EST 2012
ADH IB expression, but not ADH III, is decreased in human lung cancer.
PloS one
Mutka SC,Green LH,Verderber EL,Richards JP,Looker DL,Chlipala EA,Rosenthal GJ
Tue Jul 23 00:00:00 EDT 2013
ADH IB expression, but not ADH III, is decreased in human lung cancer.
PloS one
Mutka SC,Green LH,Verderber EL,Richards JP,Looker DL,Chlipala EA,Rosenthal GJ
Tue Jul 23 00:00:00 EDT 2013
ADH IB expression, but not ADH III, is decreased in human lung cancer.
PloS one
Mutka SC,Green LH,Verderber EL,Richards JP,Looker DL,Chlipala EA,Rosenthal GJ
Tue Jul 23 00:00:00 EDT 2013
ADH IB expression, but not ADH III, is decreased in human lung cancer.
PloS one
Mutka SC,Green LH,Verderber EL,Richards JP,Looker DL,Chlipala EA,Rosenthal GJ
Tue Jul 23 00:00:00 EDT 2013
Thioredoxin-1 regulates cellular heme insertion by controlling S-nitrosation of glyceraldehyde-3-phosphate dehydrogenase.
The Journal of biological chemistry
Chakravarti R,Stuehr DJ
Fri May 11 00:00:00 EDT 2012
Thioredoxin-1 regulates cellular heme insertion by controlling S-nitrosation of glyceraldehyde-3-phosphate dehydrogenase.
The Journal of biological chemistry
Chakravarti R,Stuehr DJ
Fri May 11 00:00:00 EDT 2012
Compartmentalized connexin 43 s-nitrosylation/denitrosylation regulates heterocellular communication in the vessel wall.
Arteriosclerosis, thrombosis, and vascular biology
Straub AC,Billaud M,Johnstone SR,Best AK,Yemen S,Dwyer ST,Looft-Wilson R,Lysiak JJ,Gaston B,Palmer L,Isakson BE
Tue Feb 01 00:00:00 EST 2011
Compartmentalized connexin 43 s-nitrosylation/denitrosylation regulates heterocellular communication in the vessel wall.
Arteriosclerosis, thrombosis, and vascular biology
Straub AC,Billaud M,Johnstone SR,Best AK,Yemen S,Dwyer ST,Looft-Wilson R,Lysiak JJ,Gaston B,Palmer L,Isakson BE
Tue Feb 01 00:00:00 EST 2011
Increased nitrosoglutathione reductase activity in hypoxic pulmonary hypertension in mice.
Journal of pharmacological sciences
Wu X,Du L,Xu X,Tan L,Li R
Mon Sep 13 00:00:00 EDT 2010
Increased nitrosoglutathione reductase activity in hypoxic pulmonary hypertension in mice.
Journal of pharmacological sciences
Wu X,Du L,Xu X,Tan L,Li R
Mon Sep 13 00:00:00 EDT 2010
Increased nitrosoglutathione reductase activity in hypoxic pulmonary hypertension in mice.
Journal of pharmacological sciences
Wu X,Du L,Xu X,Tan L,Li R
Mon Sep 13 00:00:00 EDT 2010
Increased nitrosoglutathione reductase activity in hypoxic pulmonary hypertension in mice.
Journal of pharmacological sciences
Wu X,Du L,Xu X,Tan L,Li R
Mon Sep 13 00:00:00 EDT 2010